Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA1-10815 detects Gonadotropin Releasing Hormone from human samples.
MA1-10815 has been successfully used in ELISA and immunohistochemistry applications.
The MA1-10815 immunogen is purified human gonadotropin releasing hormone.
The Luteinizing Hormone Releasing Hormone (LH-RH) is produced in the hypothalamus and influences the release of Luteinizing Hormone in the pituitary gland. LH-RH is also known as Gonadotropin-releasing hormone (Gn-RH).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GnRH-associated peptide 1; gonadotropin-releasing hormone 1 (luteinizing-releasing hormone); leuteinizing-releasing hormone; luliberin I; luteinizing hormone releasing hormone; Lutropin; Progonadoliberin I; Progonadoliberin-1; prolactin release-inhibiting factor
Gene Aliases: GNRH; GNRH1; GRH; LHRH; LNRH
UniProt ID: (Human) P01148
Entrez Gene ID: (Human) 2796
Molecular Function:
peptide hormone
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support