Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Percent identity with other species by BLAST analysis: Human, Gibbon (100%) Gorilla, Monkey, Dog, Elephant, Panda (95%) Marmoset, Bovine, Horse, Pig (90%).
This gene encodes the receptor for type 1 gonadotropin-releasing hormone. This receptor is a member of the seven-transmembrane, G-protein coupled receptor (GPCR) family. It is expressed on the surface of pituitary gonadotrope cells as well as lymphocytes, breast, ovary, and prostate. Following binding of gonadotropin-releasing hormone, the receptor associates with G-proteins that activate a phosphatidylinositol-calcium second messenger system. Activation of the receptor ultimately causes the release of gonadotropic luteinizing hormone (LH) and follicle stimulating hormone (FSH). Defects in this gene are a cause of hypogonadotropic hypogonadism (HH). Alternative splicing results in multiple transcript variants encoding different isoforms. More than 18 transcription initiation sites in the 5 region and multiple polyA signals in the 3 region have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GnRH receptor; gnRH-R; gonadotropin-releasing hormone (type 1) receptor 1; Gonadotropin-releasing hormone receptor; leutinizing hormone releasing horomone receptor; leutinizing-releasing hormone receptor; Lh-rh receptor; luliberin receptor; type I GnRH receptor; Type I GNRHR1
Gene Aliases: GNRHR; GNRHR1; GRHR; HH7; LHRHR; LRHR
UniProt ID: (Human) P30968
Entrez Gene ID: (Human) 2798
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support