View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product # 31118 has been successfully used in Western blot, IF, ICC, IHC, ELISA, and ISH applications.
Product # 31118 reacts with the heavy chains of human IgG, but does not react against non-immunoglobulin serum proteins. Based on solid-phase enzyme immunoassay, this antibody has minimal cross-reactivity with human IgE and human IgM. However, this antibody may cross-react with immunoglobulins from other species.
The activity measured by a solid phase binding assay is 91%.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.