Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Secondary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Gene Expression
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins and Peptides
  • ELISA Kits
  • SNP Genotyping
  • 所有文檔和支持
  • 證書
  • SDS(安全資料表
  • 產品常見問題
  • Documents & Support
  • 手冊和試驗方法
    Search button Close
            • 訂單查詢
            • 快速訂購
            • 登入
              登入
              還沒有帳戶嗎 ? 立即註冊
              • 帳戶
              • 訂單
              • Connect: Lab, Data, Apps
              • 客製化產品與項目
              • 儀器管理

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Secondary Antibodies ›
            • Goat anti-Mouse IgG Secondary Antibodies

            Invitrogen

            F(ab')2-Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 594

            View all (261) Goat anti-Mouse IgG antibodies
            Datasheet
            Create Citation
            Questions & Answers
            Tech Support
            Datasheet
            Create Citation
            Questions & Answers
            Tech Support

            Cite F(ab')2-Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 594

            • Antibody Testing Data (10)
            Application
            Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 10

            Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody (A48288) in ICC/IF

            Immunofluorescence analysis of F(ab)2-Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 594 (Product # A48288) was performed using SH-SY5Y (positive model) and HeLa (negative model) cells stained with Nestin Monoclonal Antibody (10C2), eBioscience™ (Product # 14-9843-80). The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 m... View More
            View Product

            Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
            Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
            Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
            Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
            Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
            Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
            Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody in Western Blot (WB)
            Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody in Western Blot (WB)

            Product Details

            A48288

            Applications
            Tested Dilution

            Western Blot (WB)

            1:2,500-1:5,000

            Immunohistochemistry (Paraffin) (IHC (P))

            1:500

            Immunocytochemistry (ICC/IF)

            1:200-1:2,000
            Product Specifications

            Species Reactivity

            Mouse

            Host/Isotype

            Goat / IgG

            Class

            Polyclonal

            Type

            Secondary Antibody

            Immunogen

            Gamma Immunoglobins Heavy and Light chains

            Conjugate

            Alexa Fluor™ Plus 594
            • Agarose
            • Alexa Fluor 350 
            • Alexa Fluor 405 
            • Alexa Fluor 430 
            • Alexa Fluor 488 
            • Alexa Fluor 514 
            • Alexa Fluor 532 
            • Alexa Fluor 546 
            • Alexa Fluor 555 
            • Alexa Fluor 568 
            • Alexa Fluor 594 
            • Alexa Fluor 633 
            • Alexa Fluor 635 
            • Alexa Fluor 647 
            • Alexa Fluor 660 
            • Alexa Fluor 680 
            • Alexa Fluor 700 
            • Alexa Fluor 750 
            • Alexa Fluor 790 
            • Alexa Fluor Plus 488 
            • Alexa Fluor Plus 555 
            • Alexa Fluor Plus 647 
            • AP
            • APC 
            • APC-Alexa Fluor 750
            • Biotin 
            • Biotin-XX
            • BODIPY FL
            • Cyanine3
            • Cyanine5 
            • DyLight 350
            • DyLight 405
            • DyLight 488
            • DyLight 550
            • DyLight 594
            • DyLight 633
            • DyLight 650
            • DyLight 680
            • DyLight 755
            • DyLight 800
            • FITC
            • HRP
            • Pacific Blue
            • Pacific Orange
            • PE 
            • PE-Alexa Fluor 610
            • Rhodamine
            • Rhodamine Red-X
            • Texas Red-X
            • TRITC
            • Request custom conjugation

            Excitation/Emission Max

            590/618 nm View spectra spectra

            Form

            Liquid

            Concentration

            2 mg/mL

            Purification

            Affinity chromatography

            Storage buffer

            proprietary buffer, pH 6.5

            Contains

            0.016% Methylisothiazolone, 0.016% Bromonitrodioxane

            Storage conditions

            4°C, store in dark

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2896354

            Target

            IgG

            Cross Adsorption

            human, horse, bovine

            Antibody Form

            F(ab')2 Fragment

            Product Specific Information

            F(ab')2 antibody fragments offer several benefits over whole IgGs, due to their smaller size (~110 kD vs. ~ 150 kD) and the absence of most of the Fc region. The smaller size allows for better penetration into tissue sections, resulting in improved staining in applications such as IHC. The lack of Fc region results in reduced nonspecific background staining when staining cells with Fc receptors (such as immune cells) and other Fc interaction-related interference.

            F(ab')2 antibody fragments were obtained by a combination of pepsin digestion and immunoaffinity chromatography to remove residual Fc fragments and undigested whole IgG molecules.

            To minimize cross-reactivity, the goat anti-mouse IgG F(ab')2 fragments have been cross-adsorbed against human, horse and bovine serum proteins. Cross-adsorption or pre-adsorption is a purification step to increase specificity of the antibody resulting in higher sensitivity and less background staining. The secondary antibody solution is passed through a column matrix containing immobilized serum proteins from potentially cross-reactive species. Only the nonspecific-binding secondary antibodies are captured in the column, and the highly specific secondaries flow through. The benefits of this extra step are apparent in multiplexing/multicolor-staining experiments (e.g., flow cytometry) where there is potential cross-reactivity with other primary antibodies or in tissue/cell fluorescent staining experiments where there may be the presence of endogenous immunoglobulins.

            Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Because staining protocols vary with application, the appropriate dilution of antibody should be determined empirically.

            Specificity: This antibody binds to heavy chains on mouse IgG and light chains on all mouse immunoglobulins. This antibody does not bind to human, horse or bovine serum proteins/IgG or mouse non-immunoglobulin serum proteins.

            Target Information

            Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            產品訂購 Plus Icon Minus Icon
            • 訂單狀態
            • 訂購協助
            • 快速訂購
            • 現貨供應中心
            • 電子採購
            支持 Plus Icon Minus Icon
            • 協助與支持
            • 聯絡我們
            • 技術支持中心
            • 搜尋文件和證書
            • 回報網站問題
            資源 Plus Icon Minus Icon
            • 學習中心
            • 促銷活動
            • 活動和網路研討會
            • 社群媒體
            關於 Thermo Fisher Plus Icon Minus Icon
            • 關於我們 關於我們
            • 人才招募 人才招募
            • 投資者資訊 投資者資訊
            • 新聞 新聞
            • 企業社會責任 企業社會責任
            • 商標
            品牌 Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Policy
            • Price & Freight Policy
            • Cookie偏好

              在此網站使用 Cookie 的選擇

              我們以及我們的附屬公司和供應商使用 cookies 和類似技術來經營我們的網站、識別我們網站的訪客、提供安全登入、收集統計資料以優化網站功能,並提供根據您的興趣量身定制的內容。點擊「全部接受」 以接受所有 cookies,並直接進入網站,或點擊「管理設定」 以查看 cookies 類型的詳細說明,並選擇是否在網站接受某些 cookies。
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            Taiwan flag icon
            Taiwan

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-vbrqw:80/100.66.75.102:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0