Search Thermo Fisher Scientific
FIGURE: 1 / 1
This antibody may cross-react with IgG1 from other species.^M
^M
Goat anti Rat IgG1 has been shown to react with rat IgG1 by immunoelectrophoresis and ELISA. Goat anti Rat IgG1 may cross react with IgG1 from other species.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.