Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Peptide sequence: LLTLALAGSS DAFSGSEATA AILSRAPWSL QSVNPGLKTN SSKEPKFTKC
Sequence homology: Horse: 83%; Human: 100%
may play a role in chondrocyte differentiation and positive regulation of longitudinal bond growth and.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GH receptor; growth hormone binding protein; Growth hormone receptor; MGC124963; MGC156665; serum binding protein; Somatotropin receptor
Gene Aliases: GHBP; GHIP; GHR
UniProt ID: (Human) P10912
Entrez Gene ID: (Human) 2690
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support