Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 16
Antibody detects endogenous levels of Acetyl-H2A.Z only when acetylated at Lys4/7/11/13.
H2A.Zac pan-acetyl (K4,K7,K11,K13) is classified under histone H2A which replaces conventional H2A in a subset of nucleosomes. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling. May be involved in the formation of constitutive heterochromatin. May be required for chromosome segregation during cell division.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H2A histone family, member Z; H2A/z; H2AZ histone; Histone H2A.Z
Gene Aliases: H2A.z; H2A.Z-1; H2A/z; H2AFZ; H2AZ; H2AZ1
UniProt ID: (Human) P0C0S5
Entrez Gene ID: (Human) 3015
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support