Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 4
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The nucleosomes wrap further and compact DNA into chromotin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones play a central role in transciption regulation, DNA repair, DNA replication, and chromosomal stability. H2B has a broad antibacterial activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acetyl-Histone H2; Acetyl-Histone H2B type 1-B K20; Acetyl-Histone H2B type 1-B Lys20; H2B GL105; H2B histone family, member F; h2B-143; H2B-clustered histone 3; H2B/f; H2B/I; H2BK12ac; H2BK15ac; H2BK20ac; H2BK5ac; H2BLys20ac; H2BS14; H2BS14p; HIST1H3I; histone 1, H2bb; Histone 2B; Histone H2B type 1-B; Histone H2B.1; Histone H2B.f; Histone H2B.I; MGC125414; MGC125415; MGC125416; MGC9388; RP1-193B12.10
Gene Aliases: H2b-143; H2B.1; H2B/f; H2BC3; H2BFF; HIST1H2BB
UniProt ID: (Human) P33778, (Mouse) Q64475
Entrez Gene ID: (Human) 3018, (Mouse) 319178
Molecular Function: histone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support