Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
This antibody is predicted to react with Mouse, Rat, Bovine.
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H3 family. Transcripts from this gene lack polyA tails; instead, they contain a palindromic termination element. This gene is located separately from the other H3 genes that are in the histone gene cluster on chromosome 6p22-p21.3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H3 histone, family 3B (H3.3B); H3.3; H3.3 K36; H3.3 K36M; H3.3K36 mutant; H3B; H3FT; H3K36M; H3t; Histone 3.3; Histone cluster 3; Histone H3.3; Histone H3.3 K36M; Histone H3.3K36M
Gene Aliases: H3-3A; H3-3B; H3.3A; H3.3B; H3F3; H3F3A; H3F3B; PP781
UniProt ID: (Human) P84243
Entrez Gene ID: (Human) 3020, (Human) 3021
Molecular Function: histone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support