Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
Anti-Histone H4 [Dimethyl Lys20] reacts with human Histone H4. A BLAST analysis was used to suggest cross-reactivity with Human, mouse, and C. elegans. Predicted to react with most mammal species. Cross-reactivity with Histone H4 from other sources has not been determined.
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CG3379-PA; CG3379-PB; CG3379-PC; CG3379-PD; di-methyl H4; di-methyl Histone H4; Di-Methyl-H4 K20; Di-Methyl-H4 Lys20; Di-Methyl-Histone H4 K20; Di-Methyl-Histone H4 Lys20; dimethyl H4; dimethyl Histone H4; H4 histone family, member A; H4 histone family, member E; H4 histone family, member I; H4 histone family, member J; H4 histone family, member M; H4 histone family, member N; H4 histone, family 2; H4 K20me2; H4K20; H4Lys20me2; His4r-PA; His4r-PB; His4r-PC; His4r-PD; HIST1H4; histone 1, H4a; histone 1, H4b; histone 1, H4e; histone 1, H4i; histone 1, H4j; histone 1, H4l; histone 2, H4; histone 2, H4a; Histone 4; Histone 4 family, member M; histone 4, H4; histone cluster 1, H4-IV, germinal H4-like 2 (similar to human histone cluster 1, class H4 genes); Histone Cluster 4; histone family member; Histone H2B 2; Histone H4; Histone H4A; Histone H4K20; Histone H4K20me2; histone IV, family 2; methyl Histone 3; methyl Histone H3; RH52884p
Gene Aliases: BcDNA:RH52884; CELE_T10C6.11; CG31611; CG3379; CG33869; CG33871; CG33873; CG33875; CG33877; CG33879; CG33881; CG33883; CG33885; CG33887; CG33889; CG33891; CG33893; CG33895; CG33897; CG33899; CG33901; CG33903; CG33905; CG33907; CG33909; dJ160A22.2; Dmel\CG3379; Dmel_CG3379; F07B7.11; F07B7.4; F45F2.12; FBtr0082962; FO108; H4; H4-12; H4-16; H4-53; H4-I; H4-II; H4-III; H4-IV; H4-V; H4-VI; H4-VII; H4/A; H4/B; H4/C; H4/D; H4/E; H4/G; H4/H; H4/I; H4/J; H4/K; H4/M; H4/N; H4/O; H4/p; H4C1; H4C11; H4C12; H4C13; H4C14; H4C15; H4C16; H4C2; H4C3; H4C4; H4C5; H4C6; H4C8; H4C9; H4f16; H4F2; H4F2iv; H4FA; H4FB; H4FC; H4FD; H4FE; H4FG; H4FH; H4FI; H4FJ; H4FK; H4FM; H4FN; H4FO; H4M; H4r; his-20; his-22; his-4; his-52; his-54; his-8; His4; His4-88CD; His4:CG31611; His4:CG33869; His4:CG33871; His4:CG33873; His4:CG33875; His4:CG33877; His4:CG33879; His4:CG33881; His4:CG33883; His4:CG33885; His4:CG33887; His4:CG33889; His4:CG33891; His4:CG33893; His4:CG33895; His4:CG33897; His4:CG33899; His4:CG33901; His4:CG33903; His4:CG33905; His4:CG33907; His4:CG33909; His4R; HIST1H4A; HIST1H4B; HIST1H4C; HIST1H4D; HIST1H4E; HIST1H4F; HIST1H4H; HIST1H4I; HIST1H4J; HIST1H4K; HIST1H4L; Hist1h4m; HIST2H4; HIST2H4A; HIST2H4B; HIST4H4; K06C4.12; K06C4.4; T10C6.11; X04652; XELAEV_18001418mg
UniProt ID: (Chicken) P62801, (Human) P62805, (Fruit fly) P84040, (Mouse) P62806, (C. elegans) Q27894, (Xenopus) P62799
Entrez Gene ID: (Chicken) 100858049, (Human) 8359, (Human) 8366, (Human) 8367, (Human) 8294, (Human) 8363, (Human) 8370, (Human) 121504, (Fruit fly) 41773, (Mouse) 326619, (Mouse) 326620, (Mouse) 319158, (Mouse) 97122, (C. elegans) 180072, (Xenopus) 447787
Molecular Function:
histone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support