Product References
Pyruvate-supported flux through medium-chain ketothiolase promotes mitochondrial lipid tolerance in cardiac and skeletal muscles.
Cell metabolism
Koves TR,Zhang GF,Davidson MT,Chaves AB,Crown SB,Johnson JM,Slentz DH,Grimsrud PA,Muoio DM
Tue Jun 06 00:00:00 UTC 2023
A coordinated multiorgan metabolic response contributes to human mitochondrial myopathy.
EMBO molecular medicine
Southwell N,Primiano G,Nadkarni V,Attarwala N,Beattie E,Miller D,Alam S,Liparulo I,Shurubor YI,Valentino ML,Carelli V,Servidei S,Gross SS,Manfredi G,Chen Q,D'Aurelio M
Mon Jul 10 00:00:00 UTC 2023
A novel isoform of hydroxyacyl-CoA dehydrogenase inhibits cell proliferation.
Biochemical and biophysical research communications
Xia G,Gao Y,Wu C,Pan H,Hou J,Su J,Wei S,Gao X
Sat May 28 00:00:00 UTC 2022
A novel isoform of hydroxyacyl-CoA dehydrogenase inhibits cell proliferation.
Biochemical and biophysical research communications
Xia G,Gao Y,Wu C,Pan H,Hou J,Su J,Wei S,Gao X
Sat May 28 00:00:00 UTC 2022
GDPD5 Related to Lipid Metabolism Is a Potential Prognostic Biomarker in Neuroblastoma.
International journal of molecular sciences
Luo T,Peng J,Li Q,Zhang Y,Huang Y,Xu L,Yang G,Tan D,Zhang Q,Tan Y
Tue Nov 08 00:00:00 UTC 2022
Quantification of Acipimox in Plasma and Tissues by LC-MS/MS: Application to Pharmacokinetic Comparison between Normoxia and Hypoxia.
Molecules (Basel, Switzerland)
Shen X,Li G,Wang L,Yu H,Zhou L,Deng H,Wang N,Lai C,Zhou W,Gao Y
Wed Sep 28 00:00:00 UTC 2022
Multi-Omics Analysis of Fatty Acid Metabolism in Thyroid Carcinoma.
Frontiers in oncology
Lu J,Zhang Y,Sun M,Ding C,Zhang L,Kong Y,Cai M,Miccoli P,Ma C,Yue X
Fri Apr 29 00:00:00 UTC 2022
SCFJFK is functionally linked to obesity and metabolic syndrome.
EMBO reports
He L,Yan R,Yang Z,Zhang Y,Liu X,Yang J,Liu X,Liu X,Xia L,Wang Y,Wu J,Wu X,Shan L,Yang X,Liang J,Shang Y,Sun L
Mon Jul 05 00:00:00 UTC 2021
Proteomic Analysis Reveals the Effect of Trichostatin A and Bone Marrow-Derived Dendritic Cells on the Fatty Acid Metabolism of NIH3T3 Cells under Oxygen-Glucose Deprivation Conditions.
Journal of proteome research
Wang X,Song T,Sun Y,Men L,Gu Y,Zhang S,Chen X
Fri Jan 01 00:00:00 UTC 2021
Effects of a Ketogenic Diet Containing Medium-Chain Triglycerides and Endurance Training on Metabolic Enzyme Adaptations in Rat Skeletal Muscle.
Nutrients
Fukazawa A,Koike A,Karasawa T,Tsutsui M,Kondo S,Terada S
Thu Apr 30 00:00:00 UTC 2020
Insulin resistance is improved in high-fat fed mice by photobiomodulation therapy at 630 nm.
Journal of biophotonics
Silva G,Ferraresi C,de Almeida RT,Motta ML,Paixão T,Ottone VO,Fonseca IA,Oliveira MX,Rocha-Vieira E,Dias-Peixoto MF,Esteves EA,Coimbra CC,Amorim FT,Magalhães FC
Sun Mar 01 00:00:00 UTC 2020
High-Intensity Interval Training Improves Markers of Oxidative Metabolism in Skeletal Muscle of Individuals With Obesity and Insulin Resistance.
Frontiers in physiology
de Matos MA,Vieira DV,Pinhal KC,Lopes JF,Dias-Peixoto MF,Pauli JR,de Castro Magalhães F,Little JP,Rocha-Vieira E,Amorim FT
Thu Oct 01 00:00:00 UTC 2020
PPARγ maintains the metabolic heterogeneity and homeostasis of renal tubules.
EBioMedicine
Lyu Z,Mao Z,Li Q,Xia Y,Liu Y,He Q,Wang Y,Zhao H,Lu Z,Zhou Q
Sat Dec 01 00:00:00 UTC 2018
PPARγ maintains the metabolic heterogeneity and homeostasis of renal tubules.
EBioMedicine
Lyu Z,Mao Z,Li Q,Xia Y,Liu Y,He Q,Wang Y,Zhao H,Lu Z,Zhou Q
Sat Dec 01 00:00:00 UTC 2018
Lack of β, β-carotene-9', 10'-oxygenase 2 leads to hepatic mitochondrial dysfunction and cellular oxidative stress in mice.
Molecular nutrition & food research
Wu L,Guo X,Hartson SD,Davis MA,He H,Medeiros DM,Wang W,Clarke SL,Lucas EA,Smith BJ,von Lintig J,Lin D
Mon May 01 00:00:00 UTC 2017
Lack of β, β-carotene-9', 10'-oxygenase 2 leads to hepatic mitochondrial dysfunction and cellular oxidative stress in mice.
Molecular nutrition & food research
Wu L,Guo X,Hartson SD,Davis MA,He H,Medeiros DM,Wang W,Clarke SL,Lucas EA,Smith BJ,von Lintig J,Lin D
Mon May 01 00:00:00 UTC 2017
Targeted Metabolomics Reveals Abnormal Hepatic Energy Metabolism by Depletion of β-Carotene Oxygenase 2 in Mice.
Scientific reports
Wu L,Guo X,Lyu Y,Clarke SL,Lucas EA,Smith BJ,Hildebrand D,Wang W,Medeiros DM,Shen X,Lin D
Tue Nov 07 00:00:00 UTC 2017
Targeted Metabolomics Reveals Abnormal Hepatic Energy Metabolism by Depletion of β-Carotene Oxygenase 2 in Mice.
Scientific reports
Wu L,Guo X,Lyu Y,Clarke SL,Lucas EA,Smith BJ,Hildebrand D,Wang W,Medeiros DM,Shen X,Lin D
Tue Nov 07 00:00:00 UTC 2017
Disruption of the histidine triad nucleotide-binding hint2 gene in mice affects glycemic control and mitochondrial function.
Hepatology (Baltimore, Md.)
Martin J,Maurhofer O,Bellance N,Benard G,Graber F,Hahn D,Galinier A,Hora C,Gupta A,Ferrand G,Hoppeler H,Rossignol R,Dufour JF,St-Pierre MV
Wed May 01 00:00:00 UTC 2013
Disruption of the histidine triad nucleotide-binding hint2 gene in mice affects glycemic control and mitochondrial function.
Hepatology (Baltimore, Md.)
Martin J,Maurhofer O,Bellance N,Benard G,Graber F,Hahn D,Galinier A,Hora C,Gupta A,Ferrand G,Hoppeler H,Rossignol R,Dufour JF,St-Pierre MV
Wed May 01 00:00:00 UTC 2013
Disruption of the histidine triad nucleotide-binding hint2 gene in mice affects glycemic control and mitochondrial function.
Hepatology (Baltimore, Md.)
Martin J,Maurhofer O,Bellance N,Benard G,Graber F,Hahn D,Galinier A,Hora C,Gupta A,Ferrand G,Hoppeler H,Rossignol R,Dufour JF,St-Pierre MV
Wed May 01 00:00:00 UTC 2013
Disruption of the histidine triad nucleotide-binding hint2 gene in mice affects glycemic control and mitochondrial function.
Hepatology (Baltimore, Md.)
Martin J,Maurhofer O,Bellance N,Benard G,Graber F,Hahn D,Galinier A,Hora C,Gupta A,Ferrand G,Hoppeler H,Rossignol R,Dufour JF,St-Pierre MV
Wed May 01 00:00:00 UTC 2013