Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: QKVEPCNDHV RAKLSWAKKR DEDDVPTVPS TLGEERLYNP FLRVAEEPVR KFTGKAVPAD VLEALCKERA RFEQAGEPRQ PQARALLALQ WGLLSAAPHD
HAGHL is a hydrolase acting on ester bonds.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GLO2-like/ RJD12; Hydroxyacylglutathione hydrolase-like protein
Gene Aliases: HAGHL
UniProt ID: (Human) Q6PII5
Entrez Gene ID: (Human) 84264
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support