Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive Samples: Mouse liver, Rat brain
Immunogen sequence: ANVTVTVLST KQTEDYCLAS NKVGRCRGSF PRWYYDPTEQ ICKSFVYGGC LGNKNNYLRE EECILACRGV QGPSMERRHP VCSGTCQPTQ FRCSNGCCID SFLECDDTPN CPDASDEAAC EKYTSGFDEL QRIHFPSDKG HCVDLPDTGL CKESIPRWYY NPFSEHCARF TYGGCYGNKN NFEEEQQCLE SCRGISKKDV FGLRREIPIP STGSVEMAVA VFLVICIVVV VAILGYCFFK NQRKDFHGHH HHPPPTPASS TVSTTEDTEH LVYNHTTRPL
The protein encoded by this gene is a member of the Kunitz family of serine protease inhibitors. The protein is a potent inhibitor specific for HGF activator and is thought to be involved in the regulation of the proteolytic activation of HGF in injured tissues. Alternative splicing results in multiple variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HAI-1; hepatocyte growth factor activator inhibitor 1; Hepatocyte growth factor activator inhibitor type 1; hepatocyte growth factor activator inhibitor-1; Kunitz-type protease inhibitor 1; serine protease inhibitor, Kunitz type 1
Gene Aliases: HAI; HAI-1; HAI1; MANSC2; SPINT1; UNQ223/PRO256
UniProt ID: (Human) O43278, (Mouse) Q9R097
Entrez Gene ID: (Human) 6692, (Mouse) 20732, (Rat) 311331
Molecular Function: protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support