Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: mouse liver.
Predicted reactivity: Mouse (96%), Rat (96%), Zebrafish (90%), Chicken (94%), Bovine (95%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HAL is a cytosolic enzyme catalyzing the first reaction in histidine catabolism, the nonoxidative deamination of L-histidine to trans-urocanic acid. HAL defects cause histidinemia which is characterized by increased histidine and histamine and decreased urocanic acid in body fluids Histidine ammonia-lyase is a cytosolic enzyme catalyzing the first reaction in histidine catabolism, the nonoxidative deamination of L-histidine to trans-urocanic acid. Histidine ammonia-lyase defects cause histidinemia which is characterized by increased histidine and histamine and decreased urocanic acid in body fluids.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Histidase; Histidine ammonia-lyase; histidinemia
Gene Aliases: HAL; HIS; histidase; Hsd; HSTD; Huth
UniProt ID: (Human) P42357, (Mouse) P35492
Entrez Gene ID: (Human) 3034, (Mouse) 15109
Molecular Function: lyase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support