Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MAERAALEEL VKLQGERVRG LKQQKASAEL IEEEVAKLLK LKAQLGPDES KQKFVLKTPK GTRDYSPRQM AVREKVFDVI IRCFKRHGAE VIDTPV
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. The protein encoded by this gene is a cytoplasmic enzyme which belongs to the class II family of aminoacyl-tRNA synthetases. The enzyme is responsible for the synthesis of histidyl-transfer RNA, which is essential for the incorporation of histidine into proteins. The gene is located in a head-to-head orientation with HARSL on chromosome five, where the homologous genes share a bidirectional promoter. The gene product is a frequent target of autoantibodies in the human autoimmune disease polymyositis/dermatomyositis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ20491; HisRS; histidine translase; Histidine--tRNA ligase, cytoplasmic; Histidyl-tRNA synthetase
Gene Aliases: CMT2W; HARS; HARS1; HRS; USH3B
UniProt ID: (Human) P12081
Entrez Gene ID: (Human) 3035
Molecular Function: aminoacyl-tRNA synthetase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support