Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Basic helix-loop-helix (bHLH) transcription factors play important roles in differentiation processes during embryonic development of vertebrates. ATOH8 (MATH6) is a tissue-restricted member of the atonal superfamily of bHLH transcription factors that exhibits 43-57% identity in the bHLH domain with other mammalian atonal paralogs including the NeuroD and Neurogenin factors. In the mouse, ATOH8 has been implicated in the specification and differentiation of neuronal cell lineages in the brain and may also participate in kidney development. Recent studies show that ATOH8 is a novel component of the pancreatic transcriptional network during embryonic development and suggest a potential role as a modulator of the differentiation program initiated by the pro-endocrine factor Neurog3. It is indispensable for early embryonic development, suggesting a more widespread function for this factor in tissue-specific differentiation processes that are dependent on class II bHLH genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: atonal homolog 6; atonal homolog 8; atonal homolog bHLH transcription factor 8; basic helix loop helix transcription factor 6; basic helix-loop-helix transcription factor 6; bHLHa21; Class A basic helix-loop-helix protein 21; hATH6; Helix-loop-helix protein hATH-6; Helix-loop-helix protein mATH-6; mATH6; Okadin; Protein atonal homolog 8; Transcription factor ATOH8
Gene Aliases: 4933425C05Rik; ATH6; ATOH8; BHLHA21; HATH6; Math6; okadin; RGD1561512
UniProt ID: (Human) Q96SQ7, (Mouse) Q99NA2
Entrez Gene ID: (Human) 84913, (Mouse) 71093, (Rat) 500200
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support