Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Peptide Sequence: RQNMDFLKAK SEEFRFGIKA AEEQLSARGM DASLSHQSLV ALSEKLARLK QQTIPLKKKL ESYLDLMPNP SLAQVKIEEA KRELDSIEAE LTRRVDMMEL
Contributes to mitotic spindle assembly, maintenance of centrosome integrity and completion of cytokinesis as part of the HAUS augmin-like complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: coiled-coil domain containing 5 (spindle associated); Coiled-coil domain-containing protein 5; Enhancer of invasion-cluster; HAUS augmin-like complex subunit 1; HAUS augmin-like complex, subunit 1; HEI-C
Gene Aliases: CCDC5; HAUS1; HEI-C; HEIC; HsT1461
UniProt ID: (Human) Q96CS2
Entrez Gene ID: (Human) 115106
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support