Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence:
MALSAEDRA LVRALWKKLG SNVGVYTTEA LERTFLAFPA TKTYFSHLDL SPGSSQVRAH GQKVADALSL AVERLDDLPH ALSALSHLHA CQLRVDPASF QLLGHCLLVT LARHYPGDFS PALQASLDKF LSHVISALVS EYR
Theta-globin mRNA is found in human fetal erythroid tissue but not in adult erythroid or other nonerythroid tissue. The theta-1 gene may be expressed very early in embryonic life, perhaps sometime before 5 weeks. Theta-1 is a member of the human alpha-globin gene cluster that involves five functional genes and two pseudogenes. The order of genes is: 5' - zeta - pseudozeta - mu - pseudoalpha-2 -pseudoalpha-1 - alpha-2 - alpha-1 - theta-1 - 3'. Research supports a transcriptionally active role for the gene and a functional role for the peptide in specific cells, possibly those of early erythroid tissue.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hemoglobin subunit theta-1; Hemoglobin theta-1 chain; hemoglobin, theta 1; Theta-1-globin
Gene Aliases: HBQ; HBQ1
UniProt ID: (Human) P09105
Entrez Gene ID: (Human) 3049
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support