Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: MAHNKIPPRW LNCPRRGQPV AGRFLPLKTM LGPRYDSQVA EENRFHPSML SNYLKSLKVK MGLLVDLTNT SRFYDRNDIE KEGIKYIKLQ CKGHGECPTT ENTETFIRLC ERFNERNPPE LIGVHCTHGF NRTGFLICAF LVEKMDWSIE AAVATFAQAR PPGIYKGDYL KELFRRYGDI EEAPPPPLLP DWCFEDDEDE DEDEDGKKES EPGSSASFGK RRKERLKLGA IFLEGVTVKG VTQVTTQPKL GEVQQKCHQF
Bifunctional mRNA-capping enzyme exhibiting RNA 5'-triphosphatase activity in the N-terminal part and mRNA guanylyltransferase activity in the C-terminal part. Catalyzes the first two steps of cap formation: by removing the gamma-phosphate from the 5'-triphosphate end of nascent mRNA to yield a diphosphate end, and by transferring the gmp moiety of GTP to the 5'-diphosphate terminus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HCAP1; HCE; MCE1; mRNA-capping enzyme; RP3-488C13.1
Gene Aliases: AU020997; CAP1A; hCAP; HCE; HCE1; MCE1; RNGTT
UniProt ID: (Human) O60942, (Mouse) O55236
Entrez Gene ID: (Human) 8732, (Mouse) 24018, (Rat) 313131
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support