Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 13
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide is homologous in mouse and marmoset.
Specificity of this antibody: HCN1.
HCN1 includes Hyperpolarization-activated cation channels of the HCN gene family, which contribute to spontaneous rhythmic activity in both heart and brain. HCN1 is a member of a family of pacemaker channels activated by hyperpolarization and regulated by cyclic nucleotides. HCN1 and HCN2 play an important role for motor learning and neuronal integration by cerebellar Purkinje cells as well as, shaping autonomous activity of single neurons and the periodicity of network oscillations. The HCN1 expression is highly enriched in cerebral cortex, hippocampus, cerebellum, and facial motor nucleus. HCN2 is highly abundant in mamillary bodies, pontine nucleus, ventral cochlear nucleus, and nucleus of the trapezoid body. These variations in regional specificity of HCN channels could generate important differences in neuronal pacemaker activity across brain systems.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BCNG-1; Brain cyclic nucleotide-gated channel 1; HAC-2; Hyperpolarization-activated cation channel 2; Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 1
Gene Aliases: BCNG-1; BCNG1; C630013B14Rik; EIEE24; HAC-2; HAC2; HCN1
UniProt ID: (Human) O60741, (Mouse) O88704
Entrez Gene ID: (Human) 348980, (Mouse) 15165
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support