Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Positive test controls include: Human coronavirus HKU1 (isolate N5) (HCoV-HKU1) Spike Protein (S1+S2 ECDHis Tag).
Immunogen sequence: NGIFSRVKNT KLYVNKTLYS EFSTIVIGSV FINNSYTIVV QPHNGVLEIT ACQYTMCEYP HTICKSKGSS RNESWHFDKS EPLCLFKKNF TYNVSTDWLY F
The S1 spike protein attaches the virion to the cell membrane by interacting with host receptor, initiating the infection. [Uniprot]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coronavirus; HCoV; HKU1; S1; Spike
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support