Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This peptide sequence is 100% homologous in multiple HDaC11 isoforms.
Suggested positive control: human, mouse, or rat brain.
This gene encodes a class IV histone deacetylase. The encoded protein is localized to the nucleus and may be involved in regulating the expression of interleukin 10. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ22237; HD11; Histone deacetylase 11
Gene Aliases: HD11; HDAC11
UniProt ID: (Human) Q96DB2, (Mouse) Q91WA3
Entrez Gene ID: (Human) 79885, (Rat) 297453, (Mouse) 232232
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support