Product References
Multi-level interaction between HIF and AHR transcriptional pathways in kidney carcinoma.
Life science alliance
Lafleur VN,Halim S,Choudhry H,Ratcliffe PJ,Mole DR
Published figure using HDAC2 polyclonal antibody (Product # PA1-861) in Western Blot
Sat Apr 01 00:00:00 UTC 2023
Multi-level interaction between HIF and AHR transcriptional pathways in kidney carcinoma.
Life science alliance
Lafleur VN,Halim S,Choudhry H,Ratcliffe PJ,Mole DR
Published figure using HDAC2 polyclonal antibody (Product # PA1-861) in Western Blot
Sat Apr 01 00:00:00 UTC 2023
Effects of Manipulation of Noradrenergic Activities on the Expression of Dopaminergic Phenotypes in Aged Rat Brains.
ASN neuro
Zeng F,Fan Y,Brown RW,Drew Gill W,Price JB,Jones TC,Zhu MY
Published figure using HDAC2 polyclonal antibody (Product # PA1-861) in Immunoprecipitation
Mon Jan 17 00:00:00 UTC 2022
Neuronal activity-induced BRG1 phosphorylation regulates enhancer activation.
Cell reports
Kim B,Luo Y,Zhan X,Zhang Z,Shi X,Yi J,Xuan Z,Wu J
PA1-861 was used in Western Blot to study how BRG1 regulates enhancer basal activities and inducibility by affecting cohesin binding, enhancer-promoter looping, RNA polymerase II recruitment, and enhancer RNA expression. Using mice with knockin mutations, to show that non-phosphorylatable BRG1 fails to efficiently induce activity-dependent genes, whereas phosphomimic BRG1 increases enhancer activity and inducibility.
Tue Jul 13 00:00:00 UTC 2021
Neuronal activity-induced BRG1 phosphorylation regulates enhancer activation.
Cell reports
Kim B,Luo Y,Zhan X,Zhang Z,Shi X,Yi J,Xuan Z,Wu J
PA1-861 was used in Western Blot to study how BRG1 regulates enhancer basal activities and inducibility by affecting cohesin binding, enhancer-promoter looping, RNA polymerase II recruitment, and enhancer RNA expression. Using mice with knockin mutations, to show that non-phosphorylatable BRG1 fails to efficiently induce activity-dependent genes, whereas phosphomimic BRG1 increases enhancer activity and inducibility.
Tue Jul 13 00:00:00 UTC 2021
Histone deacetylases meet microRNA-associated MMP-9 expression regulation in glucocorticoid-sensitive and -resistant cell lines.
International journal of oncology
Hentati-Kallel M,Le Jan S,Bernard P,Antonicelli F,Trussardi-Régnier A
PA1861 was used in western blot to suggest that the use of HDAC inhibitors in a context of acquired glucocorticoid resistance may modify the epigenetic landscape
Wed Feb 01 00:00:00 UTC 2017
Class I lysine deacetylases promote glucocorticoid-induced transcriptional repression through functional interaction with LSD1.
The Journal of steroid biochemistry and molecular biology
Patrick NM,Griggs CA,Icenogle AL,Gilpatrick MM,Kadiyala V,Jaime-Frias R,Smith CL
PA1-861 was used in Western Blotting to demonstrate that Class I lysine deacetylases facilitate dexamethasone-induced transcriptional repression by suppressing LSD1 complex activity at selected target gene promoters.
Wed Mar 01 00:00:00 UTC 2017
Loss of Asxl2 leads to myeloid malignancies in mice.
Nature communications
Li J,He F,Zhang P,Chen S,Shi H,Sun Y,Guo Y,Yang H,Man N,Greenblatt S,Li Z,Guo Z,Zhou Y,Wang L,Morey L,Williams S,Chen X,Wang QT,Nimer SD,Yu P,Wang QF,Xu M,Yang FC
PA1-861 was used in Western Blotting to demonstrate that ASXenopus laevis2 functions as a tumour suppressor to maintain normal haematopoietic stem cell function.
Thu Jun 08 00:00:00 UTC 2017
Nuclear LC3 Associates with Slowly Diffusing Complexes that Survey the Nucleolus.
Traffic (Copenhagen, Denmark)
Kraft LJ,Manral P,Dowler J,Kenworthy AK
PA1-861 was used in western blot to test nuclear LC3 and how it associates with slowly diffusing complexes that interact with the nucleolus
Fri Apr 01 00:00:00 UTC 2016
RNA-dependent dynamic histone acetylation regulates MCL1 alternative splicing.
Nucleic acids research
Khan DH,Gonzalez C,Cooper C,Sun JM,Chen HY,Healy S,Xu W,Smith KT,Workman JL,Leygue E,Davie JR
PA1-861 was used in ChIP assay, immunoprecipitation, and western blot to study the modulation of MCL1 alternative splicing by an RNA-dependent mechanism involving dynamic histone acetylation
Sat Feb 01 00:00:00 UTC 2014
Proteomic and functional analyses reveal the role of chromatin reader SFMBT1 in regulating epigenetic silencing and the myogenic gene program.
The Journal of biological chemistry
Lin S,Shen H,Li JL,Tang S,Gu Y,Chen Z,Hu C,Rice JC,Lu J,Wu L
Published figure using HDAC2 polyclonal antibody (Product # PA1-861) in Western Blot
Fri Mar 01 00:00:00 UTC 2013
Protein kinase CK2 regulates the dimerization of histone deacetylase 1 (HDAC1) and HDAC2 during mitosis.
The Journal of biological chemistry
Khan DH,He S,Yu J,Winter S,Cao W,Seiser C,Davie JR
PA1-861 was used in western blot to study the modulation of the dimerization of HDAC-1 and -2 by CK2-mediated phosphorylation during mitosis
Fri Jun 07 00:00:00 UTC 2013
Differential intranuclear organization of transcription factors Sp1 and Sp3.
Molecular biology of the cell
He S,Sun JM,Li L,Davie JR
Published figure using HDAC2 polyclonal antibody (Product # PA1-861) in Immunofluorescence
Thu Sep 01 00:00:00 UTC 2005
Differential intranuclear organization of transcription factors Sp1 and Sp3.
Molecular biology of the cell
He S,Sun JM,Li L,Davie JR
Published figure using HDAC2 polyclonal antibody (Product # PA1-861) in Immunofluorescence
Thu Sep 01 00:00:00 UTC 2005
Transcriptional repression of peroxisome proliferator-activated receptor beta/delta in murine keratinocytes by CCAAT/enhancer-binding proteins.
The Journal of biological chemistry
Di-Poï N,Desvergne B,Michalik L,Wahli W
PA1-861 was used in western blot to study the relationship between PPARbeta and C/EBP transcription factors and its effect on cell proliferation and differentiation.
Fri Nov 18 00:00:00 UTC 2005
Transcriptional repression of peroxisome proliferator-activated receptor beta/delta in murine keratinocytes by CCAAT/enhancer-binding proteins.
The Journal of biological chemistry
Di-Poï N,Desvergne B,Michalik L,Wahli W
PA1-861 was used in western blot to study the relationship between PPARbeta and C/EBP transcription factors and its effect on cell proliferation and differentiation.
Fri Nov 18 00:00:00 UTC 2005
Modification of de novo DNA methyltransferase 3a (Dnmt3a) by SUMO-1 modulates its interaction with histone deacetylases (HDACs) and its capacity to repress transcription.
Nucleic acids research
Ling Y,Sankpal UT,Robertson AK,McNally JG,Karpova T,Robertson KD
PA1-861 was used in western blot to study the regulation of DNA methyltransferase Dnmt3a enzymatic activity and transcriptional regulation.
Wed Apr 21 00:00:00 UTC 2004
Ligand-induced recruitment of a histone deacetylase in the negative-feedback regulation of the thyrotropin beta gene.
The EMBO journal
Sasaki S,Lesoon-Wood LA,Dey A,Kuwata T,Weintraub BD,Humphrey G,Yang WM,Seto E,Yen PM,Howard BH,Ozato K
PA1-861 was used in western blot to investigate the influence of histone deacetylases on the negative-feedback regulation of thyroid-stimulating hormone beta gen
Fri Oct 01 00:00:00 UTC 1999