Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation/deacetylation alters chromosome structure and affects transcription factor access to DNA. HDAC3 belongs to the histone deacetylase/acuc/apha family. It has histone deacetylase activity and represses transcription when tethered to a promoter. It may participate in the regulation of transcription through its binding with the zinc-finger transcription factor YY1. This protein can also down-regulate p53 function and thus modulate cell growth and apoptosis. This gene is regarded as a potential tumor suppressor gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CG2128-PA; HD3; Hdac3; HDAC3 splicing HDAC3alpha; HDAC3 splicing HDAC3beta; HDAC3 splicing HDAC3delta; HDAC3 splicing HDAC3gamma; HDAC3-PA; Histone deacetylase 3; histone Deacetylase-3; LD23745p; Protein deacetylase HDAC3; Protein deacylase HDAC3; RPD3-2; SMAP45
Gene Aliases: AW537363; CG2128; DHDAC3; Dmel\CG2128; Dmel_CG2128; dmHDA402; DmHDAC3; Dromel_CG2128_FBtr0078767_hdac3_mORF; HD3; HDAC; Hdac 3; HDAC3; RPD3; RPD3-2
UniProt ID: (Human) O15379, (Mouse) O88895, (Rat) Q6P6W3
Entrez Gene ID: (Fruit fly) 44446, (Human) 8841, (Mouse) 15183, (Rat) 84578
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support