Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MGFCFFNSVA IACRQLQQQS KASKILIVDW DVHHGNGTQQ TFYQDPSVLY ISLHRHDDGN FFPGSGAVDE VGAGSGEGFN VNVAWAGGLD PPMGDPEYLA AFRIVVMPIA REFSPDLVLV SAGFDAAEGH PAPLGGYHVS AKCFGYMTQQ LMNLAGGAVV LALEGGHDLT AICDASEACV AALLGNRVDP LSEEGWKQKP NLNAIRSLEA VIRVHSKYWG CMQRLASCPD SWVPRVPGAD KEEVEAVTAL ASLSVGILAE DRPSEQLVEE EEPMNL
Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation/deacetylation alters chromosome structure and affects transcription factor access to DNA. The protein encoded by this gene has sequence homology to members of the histone deacetylase family. This gene is orthologous to mouse HDAC7 gene whose protein promotes repression mediated via the transcriptional corepressor SMRT. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HD7; HD7a; hdac-7; Histone deacetylase 7; Histone deacetylase 7A; Protein deacetylase HDAC7
Gene Aliases: HD7; HD7A; HDAC7; HDAC7A
UniProt ID: (Human) Q8WUI4
Entrez Gene ID: (Human) 51564
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support