Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Papillomavirus Regulatory Factor 1/HDBP2 is a novel transcription factor shuttling between nucleus and cytoplasm and binds to the specific GCCGGCG, which is an essential cis-element for Huntington's disease gene expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HD gene regulatory region-binding protein 2; HD-regulating factor 2; HDRF-2; Huntington disease gene regulatory region-binding protein 2; Huntington's disease gene regulatory region-binding protein 2; papillomavirus regulatory factor; Papillomavirus regulatory factor 1; papillomavirus regulatory factor PRF-1; Papillomavirus-binding factor; PRF-1; Zinc finger protein 395
Gene Aliases: BC053701; Gm910; HDBP-2; HDBP2; HDRF-2; PBF; PRF-1; PRF1; Si-1-8-14; ZNF395
UniProt ID: (Human) Q9H8N7
Entrez Gene ID: (Human) 55893, (Mouse) 380912, (Rat) 305972
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support