Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalyzes the dephosphorylation of the nucleoside 5'-monophosphates deoxyadenosine monophosphate (dAMP), deoxycytidine monophosphate (dCMP), deoxyguanosine monophosphate (dGMP) and deoxythymidine monophosphate (dTMP). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5'-deoxynucleotidase HDDC2; HCV NS5A-transactivated protein 2; HD domain-containing protein 2; Hepatitis C virus NS5A-transactivated protein 2; testicular tissue protein Li 83
Gene Aliases: 2310057G13Rik; C6orf74; CGI-130; dJ167O5.2; HDDC2; NS5ATP2; RGD1307562
UniProt ID: (Human) Q7Z4H3, (Mouse) Q3SXD3
Entrez Gene ID: (Human) 51020, (Mouse) 69692, (Rat) 361462
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support