Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
1 µg/mL of MA5-45240 was sufficient for detection of HDEL-containing proteins in 10 µg of S. cerevisiae lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 78kDa.
HSP 70 family comprises four highly conserved proteins, HSP 70, HSC 70, GRP 75 and GRP 78, which serve a variety of roles. They act as molecular chaperones, facilitating the assembly of multi-protein complexes; participate in the translocation of polypeptides across cell membranes and to the nucleus; and aid in the proper folding of nascent polypeptide chains. GRP 78 is localized in the endoplasmic reticulum (ER), where it receives imported secretory proteins and is involved in the folding and translocation of nascent peptide chains. Sorting of these proteins is dependent on a C-terminal tetrapeptide signal, usually KDEL in animal cells, and HDEL in S.cerevisiae. The 2E7 clone recognizes the C-terminal peptide HDEL, a common version of the endoplasmic reticulum retention signal found in yeast, plant, nematode and other ER proteins. 2E7 specifically stains HDEL proteins in barnyard grass, beet, cotton, mung bean, sorghum and wheat.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CG5183-PA; CG5183-PB; CG5183-PC; ER lumen protein retaining receptor 2; ER lumen protein-retaining receptor; KDEL-receptor; KdelR-PA; KdelR-PB; KdelR-PC; KDELR1; lethal (2) 31Em
Gene Aliases: BEST:CK00230; CG5183; CK00230; Dmel\CG5183; Dmel_CG5183; dmErd2; ERD2; KdelR; l(2)31Em; l(2)k00311
UniProt ID: (Fruit fly) O76767
Entrez Gene ID: (Fruit fly) 34427
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support