Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
HDHD2 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory protein Li 301; Haloacid dehalogenase-like hydrolase domain-containing protein 2
Gene Aliases: 3110052N05Rik; HDHD2; HEL-S-301
UniProt ID: (Human) Q9H0R4
Entrez Gene ID: (Human) 84064
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support