Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAHRLQIRLL TWDVKDTLLR LRHPLGEAYA TKARAHGLEV EPSALEQGFR QAYRAQSHSF PNYGLSHGLT SRQWWLDVVL QTFHLAGVQD AQAVAPIAEQ LYKDFSHPCT WQVLDGAEDT LRECRTRGLR LAVISNFDRR LEGILGGLGL REHFDFVLTS EAAGWPKPDP RIFQEALRLA HMEPVVAAHV GDNYLCDYQG PRAVGMHSFL VVGPQALDPV VRDSVPKEHI LPSLAHLLPA LDCLEGSTPG L
HDHD3 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: haloacid dehalogenase-like hydrolase domain containing 3; Haloacid dehalogenase-like hydrolase domain-containing protein 3
Gene Aliases: 2810435D12Rik; C9orf158; HDHD3
UniProt ID: (Human) Q9BSH5
Entrez Gene ID: (Human) 81932
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support