Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: HeLa, HepG2, Raji, NIH-3T3, JC, BCL-1, PC-12.
Predicted reactivity: Mouse (100%), Rat (99%), Xenopus laevis (88%), Chicken (94%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
High density lipoprotein-binding protein, also known as vigilin, is a 110-kD protein that specifically binds HDL molecules and may function in the removal of excess cellular cholesterol.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HDL-binding protein; High density lipoprotein-binding protein; lipoprotein-binding protein; Vigilin
Gene Aliases: 1110005P14Rik; AA960365; AI118566; D1Ertd101e; HBP; HDLBP; PRO2900; VGL
UniProt ID: (Human) Q00341, (Mouse) Q8VDJ3
Entrez Gene ID: (Human) 3069, (Mouse) 110611, (Rat) 64474
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support