Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
HEAB is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP/GTP-binding protein; CLP1; CLP1, cleavage and polyadenylation factor I subunit, homolog; homolog of yeast CFIA subunit Clp1p; Polyadenylation factor Clp1; polyadenylation factor Clp1 {ECO:0000255|HAMAP-Rule:MF_03035}; Polynucleotide kinase Clp1; polynucleotide kinase Clp1 {ECO:0000255|HAMAP-Rule:MF_03035}; Polyribonucleotide 5'-hydroxyl-kinase Clp1; polyribonucleotide 5'-hydroxyl-kinase Clp1 {ECO:0000255|HAMAP-Rule:MF_03035}; Pre-mRNA cleavage complex II protein Clp1; pre-mRNA cleavage complex II protein Clp1 {ECO:0000255|HAMAP-Rule:MF_03035}
Gene Aliases: AI462438; CLP1; hClp1; HEAB; RGD1307679
UniProt ID: (Human) Q92989, (Mouse) Q99LI9, (Rat) Q5PQL4
Entrez Gene ID: (Human) 10978, (Mouse) 98985, (Rat) 311166
Molecular Function: mRNA polyadenylation factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support