Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 10
Product # 16842-1-AP is a rabbit polyclonal antibody raised against the full-length of human HEBP1. The MW of this protein is 21 kDa, and this antibody specifically recognizes the 21 kDa protein.
Immunogen sequence: MLGMIKNSL FGSVETWPWQ VLSKGDKEEV AYEERACEGG KFATVEVTDK PVDEALREAM PKVAKYAGGT NDKGIGMGMT VPISFAVFPN EDGSLQKKLK VWFRIPNQFQ SDPPAPSDKS VKIEEREGIT VYSMQFGGYA KEADYVAQAT RLRAALEGTA TYRGDIYFCT GYDPPMKPYG RRNEIWLLKT (1-189 aa encoded by BC016277)
The full-length protein encoded by this gene is an intracellular tetrapyrrole-binding protein. This protein includes a natural chemoattractant peptide of 21 amino acids at the N-terminus, which is a natural ligand for formyl peptide receptor-like receptor 2 (FPRL2) and promotes calcium mobilization and chemotaxis in monocytes and dendritic cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Heme-binding protein 1; p22 HBP; p22HBP
Gene Aliases: HBP; HEBP; HEBP1
UniProt ID: (Human) Q9NRV9, (Mouse) Q9R257
Entrez Gene ID: (Human) 50865, (Rat) 362454, (Mouse) 15199
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support