Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSEAVRVPSP ATPLVVAAAA PEERKGKESE REKLPPIVSA GAGATAGLDR GAKGQISTFS SFISAVSPKK EAAENRSSPA HLVFPNIKNV REPPPICLDV RQKQRTSMDA SSSEMKAPVL PEPILPIQPK TVKDFQEDVE KVKSSGDWKA VHDFYLTTFD SFPELNAAFK KDATASFNTI EDSGINAKFV NAVYDTLLNT VSIMTCK
Probable E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HECT domain containing 2; HECT domain containing E3 ubiquitin protein ligase 2; HECT domain-containing protein 2; HECT-type E3 ubiquitin transferase HECTD2; Probable E3 ubiquitin-protein ligase HECTD2
Gene Aliases: HECTD2
UniProt ID: (Human) Q5U5R9
Entrez Gene ID: (Human) 143279
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support