Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
E3 ubiquitin-protein ligase that mediates ubiquitination and subsequent degradation of DVL1. Also targets the mutant SOD1 protein involved in familial amyotrophic lateral sclerosis (FALS). Forms cytotoxic aggregates with DVL1, SSR3 and mutant SOD1 that lead to motor neuron death in FALS.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase HECW1; HECT type E3 ubiquitin ligase; HECT, C2 and WW domain-containing protein 1; HECT-type E3 ubiquitin transferase HECW1; hNEDL1; NEDD4-like E3 ubiquitin-protein ligase 1; NEDD4-like ubiquitin-protein ligase 1
Gene Aliases: HECW1; KIAA0322; NEDL1
UniProt ID: (Human) Q76N89
Entrez Gene ID: (Human) 23072
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support