Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DLECADSSCN ESVTTQLSSV DTRCSSLESA RFPETPAFSS QEEEDGACAA EPTSSGPAEG SQESVCTAGS LPVVQVPSGE DEGPGAESAT VPDQEELGEV WQRRGSLEG
E3 ubiquitin-protein ligase that mediates ubiquitination of TP73. Acts to stabilize TP73 and enhance activation of transcription by TP73.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase HECW2; hect; HECT, C2 and WW domain-containing protein 2; HECT-type E3 ubiquitin transferase HECW2; NEDD4-like E3 ubiquitin-protein ligase 2; NEDD4-related E3 ubiquitin ligase NEDL2
Gene Aliases: HECW2; KIAA1301; NEDL2
UniProt ID: (Human) Q9P2P5
Entrez Gene ID: (Human) 57520
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support