Product References
Foxp3 depends on Ikaros for control of regulatory T cell gene expression and function.
eLife
Thomas RM,Pahl MC,Wang L,Grant SFA,Hancock WW,Wells AD
Published figure using HELIOS monoclonal antibody (Product # 17-9883-42) in Flow Cytometry
Wed Apr 24 00:00:00 UTC 2024
Activation-induced cytidine deaminase expression by thymic B cells promotes T-cell tolerance and limits autoimmunity.
iScience
Lombard-Vadnais F,Chabot-Roy G,Zahn A,Rodriguez Torres S,Di Noia JM,Melichar HJ,Lesage S
Published figure using HELIOS monoclonal antibody (Product # 17-9883-42) in Flow Cytometry
Fri Jan 20 00:00:00 UTC 2023
Interaction between tissue-dwelling helminth and the gut microbiota drives mucosal immunoregulation.
NPJ biofilms and microbiomes
Wang Y,Guo A,Zou Y,Mu W,Zhang S,Shi Z,Liu Z,Cai X,Zhu XQ,Wang S
Published figure using HELIOS monoclonal antibody (Product # 17-9883-42) in Flow Cytometry
Sat Jun 24 00:00:00 UTC 2023
Dexmedetomidine alleviates acute lung injury by promoting Tregs differentiation via activation of AMPK/SIRT1 pathway.
Inflammopharmacology
Zhang ZT,Xie K,Luo RJ,Zhang DY,He ZW,Li KF,Lin SH,Xu F
Published figure using HELIOS monoclonal antibody (Product # 17-9883-42) in Flow Cytometry
Wed Feb 01 00:00:00 UTC 2023
Mesenchymal stem cells transfer mitochondria to allogeneic Tregs in an HLA-dependent manner improving their immunosuppressive activity.
Nature communications
Piekarska K,Urban-Wójciuk Z,Kurkowiak M,Pelikant-Małecka I,Schumacher A,Sakowska J,Spodnik JH,Arcimowicz Ł,Zielińska H,Tymoniuk B,Renkielska A,Siebert J,Słomińska E,Trzonkowski P,Hupp T,Marek-Trzonkowska NM
Published figure using HELIOS monoclonal antibody (Product # 17-9883-42) in Flow Cytometry
Mon Feb 14 00:00:00 UTC 2022
Notch4 signaling limits regulatory T-cell-mediated tissue repair and promotes severe lung inflammation in viral infections.
Immunity
Harb H,Benamar M,Lai PS,Contini P,Griffith JW,Crestani E,Schmitz-Abe K,Chen Q,Fong J,Marri L,Filaci G,Del Zotto G,Pishesha N,Kolifrath S,Broggi A,Ghosh S,Gelmez MY,Oktelik FB,Cetin EA,Kiykim A,Kose M,Wang Z,Cui Y,Yu XG,Li JZ,Berra L,Stephen-Victor E,Charbonnier LM,Zanoni I,Ploegh H,Deniz G,De Palma R,Chatila TA
Published figure using HELIOS monoclonal antibody (Product # 17-9883-42) in Flow Cytometry
Tue Jun 08 00:00:00 UTC 2021
Potential therapeutic effects of interleukin-35 on the differentiation of naïve T cells into Helios+Foxp3+ Tregs in clinical and experimental acute respiratory distress syndrome.
Molecular immunology
Wang C,Xie K,Li K,Lin S,Xu F
Published figure using HELIOS monoclonal antibody (Product # 17-9883-42) in Flow Cytometry
Thu Apr 01 00:00:00 UTC 2021
Adiponectin/AdipoR1 Axis Promotes IL-10 Release by Human Regulatory T Cells.
Frontiers in immunology
Ramos-Ramírez P,Malmhäll C,Tliba O,Rådinger M,Bossios A
Published figure using HELIOS monoclonal antibody (Product # 17-9883-42) in Flow Cytometry
Tue Oct 26 00:00:00 UTC 2021
Differences in Maturation Status and Immune Phenotypes of Circulating Helios+ and Helios- Tregs and Their Disrupted Correlations With Monocyte Subsets in Autoantibody-Positive T1D Individuals.
Frontiers in immunology
Zhang Y,Zhang J,Shi Y,Shen M,Lv H,Chen S,Feng Y,Chen H,Xu X,Yang T,Xu K
Published figure using HELIOS monoclonal antibody (Product # 17-9883-42) in Flow Cytometry
Wed Sep 15 00:00:00 UTC 2021
A regulatory T cell Notch4-GDF15 axis licenses tissue inflammation in asthma.
Nature immunology
Harb H,Stephen-Victor E,Crestani E,Benamar M,Massoud A,Cui Y,Charbonnier LM,Arbag S,Baris S,Cunnigham A,Leyva-Castillo JM,Geha RS,Mousavi AJ,Guennewig B,Schmitz-Abe K,Sioutas C,Phipatanakul W,Chatila TA
Published figure using HELIOS monoclonal antibody (Product # 17-9883-42) in Flow Cytometry
Sun Nov 01 00:00:00 UTC 2020
Distinct actions of the fermented beverage kefir on host behaviour, immunity and microbiome gut-brain modules in the mouse.
Microbiome
van de Wouw M,Walsh AM,Crispie F,van Leuven L,Lyte JM,Boehme M,Clarke G,Dinan TG,Cotter PD,Cryan JF
Published figure using HELIOS monoclonal antibody (Product # 17-9883-42) in Flow Cytometry
Mon May 18 00:00:00 UTC 2020
B7-CD28 co-stimulation modulates central tolerance via thymic clonal deletion and Treg generation through distinct mechanisms.
Nature communications
Watanabe M,Lu Y,Breen M,Hodes RJ
Published figure using HELIOS monoclonal antibody (Product # 17-9883-42) in Flow Cytometry
Tue Dec 08 00:00:00 UTC 2020
Glucocorticoid Receptor-Deficient Foxp3+ Regulatory T Cells Fail to Control Experimental Inflammatory Bowel Disease.
Frontiers in immunology
Rocamora-Reverte L,Tuzlak S,von Raffay L,Tisch M,Fiegl H,Drach M,Reichardt HM,Villunger A,Tischner D,Wiegers GJ
17-9883 was used in Flow cytometry/Cell sorting to generate mice with a specific deletion of the glucocorticoid receptor (GR) in Foxp3+ Treg cells.
Thu Sep 10 00:00:00 UTC 2020
IL-15-dependent balance between Foxp3 and RORγt expression impacts inflammatory bowel disease.
Nature communications
Tosiek MJ,Fiette L,El Daker S,Eberl G,Freitas AA
17-9883 was used in Flow cytometry/Cell sorting to demonstrate that impaired delivery of IL-15 to CD4+ T cells in the colon down-modulates Foxp3 expression and enhances RORγt expression.
Fri Mar 11 00:00:00 UTC 2016
Monoclonal regulatory T cells provide insights into T cell suppression.
Scientific reports
Gubser C,Schmaler M,Rossi SW,Palmer E
17-9883 was used in Flow cytometry/Cell sorting to investigate how Tregs function at a cellular and molecular level using monoclonal Tregs.
Mon May 23 00:00:00 UTC 2016
Phenotypic and functional analysis of CD1a+ dendritic cells from cats chronically infected with feline immunodeficiency virus.
Comparative immunology, microbiology and infectious diseases
Zhang L,Reckling S,Dean GA
17988342 was used in flow cytometry to investigate dendritic cell maturation and function and immune response dysregulation in feline FIV infection models
Thu Oct 01 00:00:00 UTC 2015
Human adipose-tissue derived mesenchymal stem cells induce functional de-novo regulatory T cells with methylated FOXP3 gene DNA.
Clinical and experimental immunology
Engela AU,Hoogduijn MJ,Boer K,Litjens NH,Betjes MG,Weimar W,Baan CC
17-9883 was used in Flow cytometry/Cell sorting to study whether human mesenchymal stem cell-induced CD4(+) CD25(+) CD127(-) forkhead box P3(+) T cells are functional and originate from effector T cells or represent expanded natural regulatory T cells.
Thu Aug 01 00:00:00 UTC 2013
Antigen-specific gene therapy after immunisation reduces the severity of collagen-induced arthritis.
Clinical & developmental immunology
Eneljung T,Tengvall S,Jirholt P,Henningsson L,Holmdahl R,Gustafsson K,Gjertsson I
17-9883 was used in Flow cytometry/Cell sorting to show antigen-specific gene therapy is effective when administered during the inflammatory course of arthritis.
Mon Aug 04 00:00:00 UTC 2014