Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Transcriptional repressor which binds preferentially to the canonical E box sequence 5'-CACGCG-3'.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLH transcription factor megane; Hairy and enhancer of split-related protein HELT; helt bHLH transcription factor; HES/HEY-like transcription factor; HES/HEY-like transcription factor, pseudogene 1; Heslike; Hey-like transcription factor; Hey-like transcriptional repressor; Megane bHLH factor; mesencephalic GABAergic neurons enhancer of split related protein; Protein Hes-like; Protein megane
Gene Aliases: A830086M02; bHLHb44; HCM1228; HELT; Helt-ps1; HESL; megane; Mgn; RGD1564073
UniProt ID: (Human) A6NFD8, (Mouse) Q7TS99
Entrez Gene ID: (Human) 391723, (Mouse) 234219, (Rat) 498633
Molecular Function: basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support