Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MAGENFATPF HGHVGRGAFS DVYEPAEDTF LLLDALEAAA AELAGVEICL EVGSGSGVVS AFLASMIGPQ ALYMCTDINP EAAACTLETA RCNKVHIQPV ITDLVGSHGI EAAWAGGRNG REVMDRFFPL VPDLLSPRGL FYLVTIKENN PEEILKIMKT KGLQGTTALS RQAGQETLSV LKFTKS
Heterodimeric methyltransferase that catalyzes N5-methylation of ETF1 on 'Gln-185', using S-adenosyl L-methionine as methyl donor. ETF1 needs to be complexed to ERF3 in its GTP-bound form to be efficiently methylated. May play a role in the modulation of arsenic-induced toxicity. May be involved in the conversion of monomethylarsonous acid (3+) into the less toxic dimethylarsonic acid.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HemK methyltransferase family member 2; Lysine N-methyltransferase 9; M.HsaHemK2P; Methylarsonite methyltransferase N6AMT1; Methyltransferase N6AMT1; n(6)-adenine-specific DNA methyltransferase 1; N6-DNA-methyltransferase; Protein N(5)-glutamine methyltransferase
Gene Aliases: C21orf127; HEMK2; KMT9; m.HsaHemK2P; MTQ2; N6AMT; N6AMT1; PRED28
UniProt ID: (Human) Q9Y5N5
Entrez Gene ID: (Human) 29104
Molecular Function: methyltransferase transferase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support