Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Sequence of this protein is as follows: MPADIMEKNS SSPVAASVNT TPDKPKTASE HRKSSKPIME KRRRARINES LSQLKTLILD ALKKDSSRHS KLEKADILEM TVKHLRNLQR AQMTAALSTD PSVLGKYRAG FSECMNEVTR FLSTCEGVNT EVRTRLLGHL ANCMTQINAM TYPGQPHPAL QAPPPPPPGP GGPQHAPFAP PPPLVPIPGG AAPPPGGAPC KLGSQAGEAA KVFGGFQVVP APDGQFAFLI PNGAFAHSGP VIPVYTSNSG TSVGPNAVSP SSGPSLTADS MWRPWRN
This protein belongs to the basic helix-loop-helix family of transcription factors. It is a transcriptional repressor of genes that require a bHLH protein for their transcription. The protein has a particular type of basic domain that contains a helix interrupting protein that binds to the N-box rather than the canonical E-box.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHb39; Class B basic helix-loop-helix protein 39; FLJ20408; Hairy and enhancer of split 1; Hairy homolog; Hairy-like; Hairy-like protein; hHL; Transcription factor HES-1
Gene Aliases: BHLHB39; HES-1; HES1; HHL; HL; HRY
UniProt ID: (Human) Q14469, (Mouse) P35428
Entrez Gene ID: (Human) 3280, (Mouse) 15205
Molecular Function:
basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support