Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSPSLQEGA QLGENKPSTC SFSIERILGL DQKKDCVPLM KPHRPWADTC SSSGKDGNLC LHVPNPPSGI SFPSVVDHPM PEERASKYEN YFSASERLSL KRELSWYRGR RPRTAFTQNQ IEVLENVFRV NCYPGIDIRE DLAQKLNLEE DRIQIWFQNR RAKLKRSHRE SQFLMAKKNF NTNLLE (1-185 aa encoded by B C093979)
This gene encodes a conserved homeobox protein that is a transcriptional repressor in the developing forebrain and pituitary gland. Mutations in this gene are associated with septooptic dysplasia, HESX1-related growth hormone deficiency, and combined pituitary hormone deficiency.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Anterior-restricted homeobox protein; hAnf; homeo box gene expressed in ES cells; Homeobox expressed in ES cells 1; Homeobox protein ANF; Rathke pouch homeo box; Rathke pouch homeobox
Gene Aliases: ANF; CPHD5; HANF; HES-1; HESX1; RGD1563858; RPX
UniProt ID: (Human) Q9UBX0, (Mouse) Q61658
Entrez Gene ID: (Human) 8820, (Rat) 498575, (Mouse) 15209
Molecular Function: helix-turn-helix transcription factor homeodomain transcription factor DNA-binding transcription factor gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support