FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Transcriptional regulator which functions as a general RNA polymerase II transcription inhibitor. In cooperation with 7SK snRNA sequesters P-TEFb in a large inactive 7SK snRNP complex preventing RNA polymerase II phosphorylation and subsequent transcriptional elongation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hexamethylene bis-acetamide inducible 2; Hexamethylene bis-acetamide-inducible protein 2; hexamethylene-bis-acetamide-inducible transcript 2; MAQ1 paralog; Protein HEXIM2
Gene Aliases: HEXIM2; L3
UniProt ID: (Human) Q96MH2
Entrez Gene ID: (Human) 124790
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support