Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PHVALYALEV MESVVKNCGQ TVHDEVANKQ TMEELKDLLK RQVEVNVRNK ILYLIQAWAH AFRNEPKYKV VQDTYQIMKV EGHVFPEFKE SDAMFAAERA PDWVDAEECH RCRVQFGVMT RKHHCRACGQ IFCGKCS
HGS is involved in intracellular signal transduction mediated by cytokines and growth factors. When associated with STAM, it suppresses DNA signaling upon stimulation by IL-2 and GM-CSF. It could be a direct effector of PI3-kinase in vesicular pathway via early endosomes and may regulate trafficking to early and late endosomes by recruiting clathrin. HGS may concentrate ubiquitinated receptors within clathrin-coated regions. It is involved in down-regulation of receptor tyrosine kinase via multivesicular body (MVBs) when complexed with STAM. This complex binds ubiquitin and acts as sorting machinery that recognizes ubiquitinated receptors and transfers them to further sequential lysosomal sorting/trafficking processes. HGS may contribute to the efficient recruitment of SMADs to the activin receptor complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hepatocyte growth factor-regulated tyrosine kinase substrate; HGNC:4897; Hrs; human growth factor-regulated tyrosine kinase substrate; Protein pp110
Gene Aliases: HGS; HRS
UniProt ID: (Human) O14964
Entrez Gene ID: (Human) 9146
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support