Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
HHIPL2 gene ontology annotations related to this gene include catalytic activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hedgehog interacting protein-like 2; HHIP-like 2; HHIP-like protein 2
Gene Aliases: HHIP3; HHIPL2; KIAA1822L; UNQ841/PRO1779
UniProt ID: (Human) Q6UWX4
Entrez Gene ID: (Human) 79802
Molecular Function:
protein-binding activity modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support