Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGQREMWRL MSRFNAFKRT NTILHHLRMS KHTDAAEEVL LGKKGCTGVI TLNRPKFLNA LTLNMIRQIY PQLKKWEQDP ETFLIIIKGA GGKAFCAGGD IRVISEAEKA KQKIAPVFFR EEYMLNNAVG SCQKPYVALI HGITMGGGVG LSVHGQFRVA TEKCLFAMPE TAIGLFPDVG GGYFLPRLQG KLGYFLALTG FRLKGRDVYR AGIATHFVDS EKLAMLEEDL LALKSPSKEN IASVLENYHT ESKIDRDKSF ILEEHMDKIN SCFSANTVEE IIENLQQDGS SFALEQLKVI NKMSPTSLKI TLRQLMEGSS KTLQEVLTME YRLSQACMRG HDFHEGVRAV LIDKDQSPKW KPADLKEVTE EDLNNHFKSL GSSDLKF (1-386 aa encoded by BC067822)
Beta-hydroxyisobutyryl-CoA hydrolase (EC 3.1.2.4) is responsible for the specific hydrolysis of HIBYL-CoA, a valine catabolite, as well as the hydrolysis of beta-hydroxypropionyl-CoA, an intermediate in a minor pathway of propionate metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-hydroxyisobutyryl-CoA hydrolase, mitochondrial; 3-hydroxyisobutyryl-Coenzyme A hydrolase; HIB-CoA hydrolase; HIBYL-COA-H; testicular tissue protein Li 86
Gene Aliases: 2610509I15Rik; AI648812; HIBCH; HIBYLCOAH
UniProt ID: (Human) Q6NVY1, (Rat) Q5XIE6, (Mouse) Q8QZS1
Entrez Gene ID: (Human) 26275, (Rat) 301384, (Mouse) 227095
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support