Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant mouse HIN-1 is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
HIN-1, also known as High In Normal-1 or Secretoglobin Family 3A Member 1 (SCGB3A1), is a gene located on chromosome 5q33.3. It encodes a protein that is predominantly expressed in normal mammary epithelial cells and is thought to function in epithelial cell signaling, contributing to cellular differentiation and maintaining normal tissue architecture. The HIN-1 gene is often silenced or downregulated in various cancers, particularly breast cancer, due to hypermethylation of its promoter region, suggesting a potential tumor suppressor role. Loss of HIN-1 expression has been implicated in the progression of malignancies by disrupting normal cell signaling pathways, thereby facilitating oncogenesis. Given its significant downregulation in cancerous tissues, HIN-1 is being explored as a biomarker for early cancer detection and may provide insights into novel therapeutic strategies that aim to restore its expression and normal cellular functions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cytokine high in normal-1; Cytokine HIN-1; High in normal 1; Pneumo secretory protein 2; PnSP-2; Secretoglobin family 3A member 1; secretoglobin, family 3A, member 1; UGRP2MGC87867; Uteroglobin-related protein 2
Gene Aliases: HIN-1; HIN1; LU105; PnSP-2; PNSP2; SCGB3A1; UGRP2; UNQ629/PRO1245
UniProt ID: (Human) Q96QR1
Entrez Gene ID: (Human) 92304
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support