Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RASDMLYFK RLIQIPRLPE GPPNFLRASA LAEHIKPVVV IPEEAPEDEE PENLIEISTG PPAGEPVVVA DLFDQTFGPP SGSVKDDRDL QIESLKREVE MLRSELEKIK LEAQRYIAQL KSQVNALEGE LEEQRKQKQK ALVDNEQLRH ELAQLRAAQL EGERSQGLRE EAERKASATE ARYNKLKEKH SELVHVHAEL LRKNADTAKQ LTVTQQSQEE VARVKEQLAF QVEQVKRESE LKLEEKSDQL EKLKRELEAK AGELARAQEA LSHTEQSKSE LSSRLDTLSA EKDALSGAVR QREADLLAAQ SLVRETEAAL SREQQRSSQE QGELQGRLAE RVWPPQMQQH H (266-615 aa encoded by BC067085)
Huntingtin disease, a neurodegenerate disease, is caused by the expansion of a polymorphic glutamine tract in huntingtin. The Huntingtin Interacting Protein 1 (HIP-1) is a reportedly proapoptotic, cargo-specific adaptor protein that may be involved in the pathogenesis of Huntingtin disease. As well as playing a role in Huntingtin disease, it is likely to be involved in the recruitment of clathrin coats to lipid membranes and it may also factor in tumorigenesis by allowing the survival of precancerous and cancerous cells. Since HIP-1 expression is significantly associated with prostate and colon cancer metastasis, HIP-1 can serve as a putative prognostic factor for prostate and colon cancers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HIP-12; HIP1-related protein; huntingtin interacting protein 12; Huntingtin-interacting protein 1-related protein; Huntingtin-interacting protein 12
Gene Aliases: AA410023; HIP12; HIP1R; HIP3; ILWEQ; KIAA0655; mKIAA0655
UniProt ID: (Human) O75146, (Mouse) Q9JKY5
Entrez Gene ID: (Human) 9026, (Mouse) 29816, (Rat) 81917
Molecular Function:
non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support