Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Fabgennix
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
HIPK3 is a serine/threonine kinase involved in signal transduction, cellular communication, and multidrug resistance of tumors and plays a role in the phosphorylation of FADD. HIPK3 negatively regulates apoptosis by promoting FADD phosphorylation. This kinase enhances androgen receptor-mediated transcription, and may act as a transcriptional corepressor for NK homeodomain transcription factors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Androgen receptor-interacting nuclear protein kinase; ANPK; Fas-interacting serine/threonine-protein kinase; FIST; HCMV-interacting protein; Homeodomain-interacting protein kinase 3; Homolog of protein kinase YAK1; Nbak3; Nuclear body-associated kinase 3; nuclear serine/threonine protein kinase; RP1-8L15.1
Gene Aliases: ANPK; DYRK6; Fist; FIST3; HIPK3; mir-1902; PKY; YAK1
UniProt ID: (Human) Q9H422, (Mouse) Q9ERH7, (Rat) O88850
Entrez Gene ID: (Human) 10114, (Mouse) 15259, (Rat) 83617
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support