Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MWSLTASEGE STTAHFFLGA GDEGLGTRGI GMRPEESDSE LLEDEEDEVP PEPQIIVGIC AMTKKSKSKP MTQILERLCR FDYLTVVILG EDVILNEPVE NWPSCHCLIS FHSKGFPLDK AVAYSKLRNP FLINDLAMQY YIQDRREVYR ILQEEGIDLP RYAVLNRDPA RPEECNLIEG EDQVEVNGAV FPKPFVEKPV SAEDHNVYIY YPSSAGGGSQ RLFRKIGSRS SVYSPESSVR KTGSYIYEEF MPTDGTDVKV YTVGPDYAHA EARKSPALDG KVERDSEGKE IRYPVMLTAM EKLVARKVCV AFKQTVCGFD LLRANGHSFV CDVNGFSFVK NSMKYYDDCA KILGNTIMRE LAPQFQIPWS IPTEAEDIPI VPTTSGTMME LRCVIAIIRH GDRTPKQKMK MEVKHPRFFA LFEKHGGYKT GKLKLKRPEQ LQEVLDITRL LLAELEKEPG GEIEEKTGKL EQLKSVLEMY GHFSGINRKV QLTYYPHGVK ASNEGQDPQR ETLAPSLLLV LKWGGELTPA GRVQAEELGR AFRCMYPGGQ GDYAGFPGCG LLRLHSTFRH DLKIYASDEG RVQMTAAAFA KGLLALEGEL TPILVQMVKS ANMNGLLDSD GDSLSSCQHR VKARLHHILQ QDAPFGPEDY DQLAPTRSTS LLNSMTIIQN PVKVCDQVFA LIENLTHQIR ERMQDPRSVD LQLYHSETLE LMLQRWSKLE RDFRQKSGRY DISKIPDIYD CVKYDVQHNG SLGLQGTAEL LRLSKALADV VIPQEYGISR EEKLEIAVGF CLPLLRKILL DLQRTHEDES VNKLHPLCYL RYSRGVLSPG RHVRTRLYFT SESHVHSLLS VFRYGGLLDE TQDAQWQRAL DYLSAISELN YMTQIVIMLY EDNTQDPLSE ERFHVELHFS PGVKGVEEEG SAPAGCGFRP ASSENEEMKT NQGSMENLCP GKASDEPDRA LQTSPQPPEG PGLPRRSPLI RNRKAGSMEV LSETSSSRPG GYRLFSSSRP PTEMKQSGLG SQCTGLFSTT VLGGSSSAPN LQDYARSHGK KLPPASLKHR DELLFVPAVK RFSVSFAKHP TNGFEGCSMV PTIYPLETLH NALSLRQVSE FLSRVCQRHT DAQAQASAAL FDSMHSSQAS DNPFSPPRTL HSPPLQLQQR SEKPPWYSSG PSSTVSSAGP SSPTTVDGNS QFGFSDQPSL NSHVAEEHQG LGLLQETPGS GAQELSIEGE QELFEPNQSP QVPPMETSQP YEEVSQPCQE VPDISQPCQD ISEALSQPCQ KVPDISQQCQ ENHDNGNHTC QEVPHISQPC QKSSQLCQKV SEEVCQLCLE NSEEVSQPCQ GVSVEVGKLV HKFHVGVGSL VQETLVEVGS PAEEIPEEVI QPYQEFSVEV GRLAQETSAI NLLSQGIPEI DKPSQEFPEE IDLQAQEVPE EIN
This gene encodes a dual functional inositol kinase. The encoded enzyme converts inositol hexakisphosphate to diphosphoinositol pentakisphosphate and diphosphoinositol pentakisphosphate to bis-diphosphoinositol tetrakisphosphate. This protein may be important for intracellular signaling pathways. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 15.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Diphosphoinositol pentakisphosphate kinase 1; histidine acid phosphatase domain containing 2A; Histidine acid phosphatase domain-containing protein 2A; hsVIP1; Inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 1; Inositol pyrophosphate synthase 1; inositol-hexakisphosphate kinase; InsP6 and PP-IP5 kinase 1; IP6 kinase; VIP1 homolog
Gene Aliases: HISPPD2A; hsVIP1; IP6K; IPS1; KIAA0377; PPIP5K1; VIP1
UniProt ID: (Human) Q6PFW1
Entrez Gene ID: (Human) 9677
Molecular Function:
nucleotide kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support