Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H2B histone family, member F; H2B-clustered histone 3; H2B/f; H2BFF; HIST1H2BB; histone 1, H2bb; Histone H2B type 1-B; Histone H2B.1; Histone H2B.f
Gene Aliases: H2B.1; H2B/f; H2BC3; H2BFF; HIST1H2BB
UniProt ID: (Human) P33778
Entrez Gene ID: (Human) 3018
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support